Assay Design
Gene Transcripts
- Gene Symbol
- lgc-9
- Entrez Gene ID
-
182195
- Gene Name
- Ligand-Gated ion Channel
- Gene Aliases
-
CELE_C04C3.2
- Chromosome Location
-
Chr.IV: 3415683 - 3419234 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067940.5 |
NP_500341.1 |
4 - 5 |
634 |
67 |
View Full Product Details