Assay Design
Gene Transcripts
- Gene Symbol
- aat-8
- Entrez Gene ID
-
185079
- Gene Name
- Amino Acid Transporter
- Gene Aliases
-
CELE_F28F9.4
- Chromosome Location
-
Chr.IV: 3862033 - 3865044 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068024.3 |
NP_500425.2 |
7 - 8 |
1063 |
65 |
View Full Product Details