Assay Design
Gene Transcripts
- Gene Symbol
- oac-23
- Entrez Gene ID
-
185419
- Gene Name
- Acyl_transf_3 domain-containing protein
- Gene Aliases
-
CELE_F37C4.3
- Chromosome Location
-
Chr.IV: 3881441 - 3884688 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068030.2 |
NP_500431.1 |
11 - 12 |
1874 |
96 |
View Full Product Details