Assay Design
Gene Transcripts
- Gene Symbol
- clec-71
- Entrez Gene ID
-
189945
- Gene Name
- C-type lectin domain-containing protein
- Gene Aliases
-
CELE_Y46C8AL.4
- Chromosome Location
-
Chr.IV: 3937458 - 3939979 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068039.4 |
NP_500440.2 |
3 - 4 |
689 |
70 |
View Full Product Details