Assay Design
Gene Transcripts
- Gene Symbol
- clec-78
- Entrez Gene ID
-
185915
- Gene Name
- C-type LECtin
- Gene Aliases
-
CELE_F47C12.2
- Chromosome Location
-
Chr.IV: 3989102 - 4014235 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068053.5 |
NP_500454.2 |
2 - 3 |
391 |
73 |
View Full Product Details