Assay Design
Gene Transcripts
- Gene Symbol
- rnp-2
- Entrez Gene ID
-
177181
- Gene Name
- RNP (RRM RNA binding domain) containing
- Gene Aliases
-
CELE_K08D10.4
- Chromosome Location
-
Chr.IV: 4172076 - 4174016 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068103.5 |
NP_500504.1 |
1 - 2 |
213 |
118 |
View Full Product Details