Assay Design
Gene Transcripts
- Gene Symbol
- fbxa-86
- Entrez Gene ID
-
189417
- Gene Name
- F-box domain-containing protein
- Gene Aliases
-
CELE_Y9C9A.12
- Chromosome Location
-
Chr.IV: 4936647 - 4938013 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068277.3 |
NP_500678.2 |
1 - 2 |
71 |
84 |
View Full Product Details