Assay Design
Gene Transcripts
- Gene Symbol
- htp-1
- Entrez Gene ID
-
259571
- Gene Name
- HORMA domain-containing protein
- Gene Aliases
-
CELE_F41H10.10
- Chromosome Location
-
Chr.IV: 5390184 - 5393660 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068398.4 |
NP_500799.2 |
3 - 4 |
282 |
81 |
View Full Product Details