Assay Design
Gene Transcripts
- Gene Symbol
- srsx-14
- Entrez Gene ID
-
183350
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_C39H7.5
- Chromosome Location
-
Chr.IV: 5624855 - 5627156 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068434.2 |
NP_500835.2 |
3 - 4 |
470 |
126 |
View Full Product Details