Assay Design
Gene Transcripts
- Gene Symbol
- nhr-46
- Entrez Gene ID
-
177350
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C45E5.6
- Chromosome Location
-
Chr.IV: 5747363 - 5757862 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068457.6 |
NP_500858.2 |
1 - 2 |
287 |
61 |
View Full Product Details