Assay Design
Gene Transcripts
- Gene Symbol
- srv-22
- Entrez Gene ID
-
186687
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_H04M03.9
- Chromosome Location
-
Chr.IV: 5878470 - 5879953 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068480.1 |
NP_500881.1 |
5 - 6 |
677 |
144 |
View Full Product Details