Assay Design



Gene Transcripts

Gene Symbol
kcc-2
Entrez Gene ID
177370
Gene Name
protein-coding
Gene Aliases
CELE_H16O14.1
Chromosome Location
Chr.IV: 6040976 - 6074507 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268349.2 NP_001255278.1 1 - 2 263 97
GenBank mRNA FJ593901.1 - 1 - 2 132 97

View Full Product Details