Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001268349.2 | NP_001255278.1 | 13 - 14 | 3285 | 68 |
NM_001268350.4 | NP_001255279.2 | 14 - 15 | 3304 | 68 | |
NM_001268351.3 | NP_001255280.1 | 13 - 14 | 3499 | 68 | |
NM_001268352.3 | NP_001255281.1 | 13 - 14 | 3100 | 68 | |
NM_001380258.1 | NP_001368424.1 | 11 - 12 | 2932 | 68 | |
GenBank mRNA | FJ593901.1 | - | 13 - 14 | 3154 | 68 |
FJ593903.1 | - | 13 - 14 | 3100 | 68 |