Assay Design



Gene Transcripts

Gene Symbol
aat-4
Entrez Gene ID
177388
Gene Name
Amino Acid Transporter
Gene Aliases
CELE_T13A10.10
Chromosome Location
Chr.IV: 6272529 - 6275721 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028211.4 NP_001023382.1 7 - 8 971 134
NR_131543.1 7 - 8 980 134

View Full Product Details