Assay Design
Gene Transcripts
- Gene Symbol
- ops-1
- Entrez Gene ID
-
188465
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_T13A10.13
- Chromosome Location
-
Chr.IV: 6282913 - 6284387 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068555.3 |
NP_500956.1 |
2 - 3 |
404 |
149 |
View Full Product Details