Assay Design
Gene Transcripts
- Gene Symbol
- srv-24
- Entrez Gene ID
-
190646
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_Y73B6BL.10
- Chromosome Location
-
Chr.IV: 6288364 - 6290841 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068556.1 |
NP_500957.1 |
4 - 5 |
495 |
80 |
View Full Product Details