Assay Design



Gene Transcripts

Gene Symbol
seu-1
Entrez Gene ID
177394
Gene Name
protein-coding
Gene Aliases
CELE_Y73B6BL.5
Chromosome Location
Chr.IV: 6329639 - 6333565 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068564.7 NP_500965.1 1 - 2 262 62
NM_171359.5 NP_741421.2 1 - 2 262 62

View Full Product Details