Assay Design
Gene Transcripts
- Gene Symbol
- tag-89
- Entrez Gene ID
-
178062
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_H02I12.3
- Chromosome Location
-
Chr.IV: 11394543 - 11396818 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069746.2 |
NP_502147.1 |
1 - 2 |
181 |
80 |
View Full Product Details