Assay Design
Gene Transcripts
- Gene Symbol
- wht-2
- Entrez Gene ID
-
178069
- Gene Name
- ABC transporter domain-containing protein
- Gene Aliases
-
CELE_C10C6.5
- Chromosome Location
-
Chr.IV: 11470436 - 11473422 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069763.7 |
NP_502164.1 |
8 - 9 |
1131 |
136 |
View Full Product Details