Assay Design



Gene Transcripts

Gene Symbol
sms-1
Entrez Gene ID
178072
Gene Name
protein-coding
Gene Aliases
CELE_H21P03.3
Chromosome Location
Chr.IV: 11482662 - 11502094 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136389.4 NP_001129861.2 3 - 4 700 68
NM_001268630.3 NP_001255559.1 3 - 4 889 68
NM_069767.5 NP_502168.2 6 - 7 1121 68
NM_069768.6 NP_502169.2 4 - 5 1022 68

View Full Product Details