Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001268663.2 | NP_001255592.1 | 5 - 6 | 721 | 96 |
NM_001268664.3 | NP_001255593.1 | 4 - 5 | 765 | 96 | |
NR_131613.1 | 5 - 6 | 720 | 96 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001380473.1 | NP_001366838.1 | 5 - 6 | 751 | 96 |