Assay Design



Gene Transcripts

Gene Symbol
nhr-43
Entrez Gene ID
178121
Gene Name
protein-coding
Gene Aliases
CELE_C29E6.5
Chromosome Location
Chr.IV: 11899590 - 11902976 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307047.3 NP_001293976.1 4 - 5 497 71
NM_069853.5 NP_502254.1 4 - 5 432 71

View Full Product Details