Assay Design
Gene Transcripts
- Gene Symbol
- lips-6
- Entrez Gene ID
-
191364
- Gene Name
- LIPaSe related
- Gene Aliases
-
CELE_ZK617.2
- Chromosome Location
-
Chr.IV: 12014128 - 12015686 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069875.3 |
NP_502276.1 |
2 - 3 |
390 |
73 |
View Full Product Details