Assay Design



Gene Transcripts

Gene Symbol
lin-24
Entrez Gene ID
178159
Gene Name
protein-coding
Gene Aliases
CELE_B0001.1
Chromosome Location
Chr.IV: 12129504 - 12135096 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268695.2 NP_001255624.1 3 - 4 363 63
NM_001268696.3 NP_001255625.1 1 - 2 42 63

View Full Product Details