Assay Design
Gene Transcripts
- Gene Symbol
- nlr-1
- Entrez Gene ID
-
178167
- Gene Name
- Neurexin Like receptor
- Gene Aliases
-
CELE_F20B10.1
- Chromosome Location
-
Chr.IV: 12170824 - 12186605 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069911.5 |
NP_502312.2 |
18 - 19 |
3950 |
106 |
View Full Product Details