Assay Design
Gene Transcripts
- Gene Symbol
- dpm-3
- Entrez Gene ID
-
178189
- Gene Name
- Probable dolichol-phosphate mannosyltransferase subunit 3
- Gene Aliases
-
CELE_F28D1.11
- Chromosome Location
-
Chr.IV: 12391587 - 12392120 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069965.4 |
NP_502366.1 |
2 - 3 |
166 |
96 |
View Full Product Details