Assay Design
Gene Transcripts
- Gene Symbol
- gex-3
- Entrez Gene ID
-
178191
- Gene Name
- Membrane-associated protein gex-3
- Gene Aliases
-
CELE_F28D1.10
- Chromosome Location
-
Chr.IV: 12404431 - 12408697 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069967.6 |
NP_502368.1 |
10 - 11 |
3098 |
78 |
GenBank mRNA |
AB073210.1 |
- |
10 - 11 |
3094 |
78 |
View Full Product Details