Assay Design
Gene Transcripts
- Gene Symbol
- him-6
- Entrez Gene ID
-
178201
- Gene Name
- Bloom syndrome protein homolog
- Gene Aliases
-
CELE_T04A11.6
- Chromosome Location
-
Chr.IV: 12490791 - 12495298 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069989.7 |
NP_502390.2 |
12 - 13 |
2838 |
85 |
GenBank mRNA |
AY095296.1 |
- |
12 - 13 |
2860 |
85 |
View Full Product Details