Assay Design
Gene Transcripts
- Gene Symbol
- sru-13
- Entrez Gene ID
-
186151
- Gene Name
- Serpentine Receptor, class U
- Gene Aliases
-
CELE_F53B2.4
- Chromosome Location
-
Chr.IV: 12521239 - 12522621 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069996.2 |
NP_502397.2 |
3 - 4 |
590 |
102 |
View Full Product Details