Assay Design
Gene Transcripts
- Gene Symbol
- rbd-1
- Entrez Gene ID
-
178229
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_T23F6.4
- Chromosome Location
-
Chr.IV: 12724986 - 12730100 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070031.5 |
NP_502432.1 |
3 - 4 |
1326 |
95 |
View Full Product Details