Assay Design



Gene Transcripts

Gene Symbol
rbd-1
Entrez Gene ID
178229
Gene Name
protein-coding
Gene Aliases
CELE_T23F6.4
Chromosome Location
Chr.IV: 12724986 - 12730100 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306991.3 NP_001293920.1 1 - 2 527 67
NM_070031.5 NP_502432.1 4 - 5 1983 67

View Full Product Details