Assay Design
Gene Transcripts
- Gene Symbol
- flp-9
- Entrez Gene ID
-
178232
- Gene Name
- KPSFVRF-amide 2
- Gene Aliases
-
CELE_C36H8.3
- Chromosome Location
-
Chr.IV: 12754558 - 12757673 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070035.7 |
NP_502436.2 |
1 - 2 |
237 |
86 |
View Full Product Details