Assay Design
Gene Transcripts
- Gene Symbol
- gcy-18
- Entrez Gene ID
-
178237
- Gene Name
- Receptor-type guanylate cyclase gcy-18
- Gene Aliases
-
CELE_ZK896.8
- Chromosome Location
-
Chr.IV: 12860097 - 12865956 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070048.7 |
NP_502449.2 |
4 - 5 |
605 |
65 |
GenBank mRNA |
AB201389.1 |
- |
4 - 5 |
565 |
65 |
View Full Product Details