Assay Design
Gene Transcripts
- Gene Symbol
- dct-19
- Entrez Gene ID
-
183136
- Gene Name
- CUB_2 domain-containing protein
- Gene Aliases
-
CELE_C32H11.13
- Chromosome Location
-
Chr.IV: 12942933 - 12944526 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070074.4 |
NP_502475.2 |
3 - 4 |
397 |
88 |
View Full Product Details