Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001026105.4 | NP_001021276.1 | 4 - 5 | 707 | 64 |
NM_001026106.2 | NP_001021277.1 | 4 - 5 | 594 | 64 | |
NM_001268771.3 | NP_001255700.1 | 1 - 2 | 56 | 64 | |
NM_001268772.4 | NP_001255701.1 | 4 - 5 | 439 | 64 |