Assay Design



Gene Transcripts

Gene Symbol
ced-3
Entrez Gene ID
178272
Gene Name
protein-coding
Gene Aliases
CELE_C48D1.2
Chromosome Location
Chr.IV: 13198261 - 13204137 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268779.3 NP_001255708.1 4 - 5 941 69
NM_001268780.3 NP_001255709.1 1 - 2 222 69
GenBank mRNA AF210702.1 - 4 - 5 885 69

View Full Product Details