Assay Design
Gene Transcripts
- Gene Symbol
- cho-1
- Entrez Gene ID
-
178274
- Gene Name
- High-affinity choline transporter 1
- Gene Aliases
-
CELE_C48D1.3
- Chromosome Location
-
Chr.IV: 13209729 - 13214566 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070138.8 |
NP_502539.1 |
1 - 2 |
247 |
80 |
GenBank mRNA |
AB030946.1 |
- |
1 - 2 |
248 |
80 |
View Full Product Details