Assay Design
Gene Transcripts
- Gene Symbol
- mex-5
- Entrez Gene ID
-
178296
- Gene Name
- Zinc finger protein mex-5
- Gene Aliases
-
CELE_W02A2.7
- Chromosome Location
-
Chr.IV: 13353716 - 13356104 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070165.6 |
NP_502566.1 |
3 - 4 |
1075 |
70 |
View Full Product Details