Assay Design



Gene Transcripts

Gene Symbol
tbc-9
Entrez Gene ID
178316
Gene Name
protein-coding
Gene Aliases
CELE_Y45F10A.6
Chromosome Location
Chr.IV: 13476932 - 13502856 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129340.5 NP_001122812.1 5 - 6 625 136
NM_001307025.4 NP_001293954.1 5 - 6 625 136
NM_070197.6 NP_502598.2 5 - 6 625 136
NM_070198.5 NP_502599.2 5 - 6 619 136

View Full Product Details