Assay Design



Gene Transcripts

Gene Symbol
tbc-9
Entrez Gene ID
178316
Gene Name
protein-coding
Gene Aliases
CELE_Y45F10A.6
Chromosome Location
Chr.IV: 13476932 - 13502856 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129340.5 NP_001122812.1 8 - 9 1044 81
NM_001276877.3 NP_001263806.1 4 - 5 466 81
NM_001276878.3 NP_001263807.1 4 - 5 466 81
NM_001276879.3 NP_001263808.1 4 - 5 466 81
NM_001307025.4 NP_001293954.1 8 - 9 1044 81
NM_070197.6 NP_502598.2 8 - 9 1044 81
NM_070198.5 NP_502599.2 8 - 9 1038 81

View Full Product Details