Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001047487.3 | NP_001040952.2 | 18 - 19 | 3122 | 136 |
NM_001047488.4 | NP_001040953.1 | 15 - 16 | 2552 | 136 | |
NM_001268820.3 | NP_001255749.1 | 11 - 12 | 1778 | 136 | |
NM_001268821.3 | NP_001255750.1 | 3 - 4 | 416 | 136 |