Assay Design



Gene Transcripts

Gene Symbol
rsd-2
Entrez Gene ID
178322
Gene Name
protein-coding
Gene Aliases
CELE_F52G2.2
Chromosome Location
Chr.IV: 13521970 - 13540265 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047487.3 NP_001040952.2 10 - 11 2007 104
NM_001047488.4 NP_001040953.1 7 - 8 1437 104
NM_001268820.3 NP_001255749.1 3 - 4 663 104

View Full Product Details