Assay Design
Gene Transcripts
- Gene Symbol
- sru-12
- Entrez Gene ID
-
189916
- Gene Name
- Serpentine Receptor, class U
- Gene Aliases
-
CELE_Y45F10B.5
- Chromosome Location
-
Chr.IV: 13583209 - 13584479 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070216.6 |
NP_502617.2 |
3 - 4 |
612 |
116 |
View Full Product Details