Assay Design
Gene Transcripts
- Gene Symbol
- oac-48
- Entrez Gene ID
-
188814
- Gene Name
- Acyl_transf_3 domain-containing protein
- Gene Aliases
-
CELE_T23G4.4
- Chromosome Location
-
Chr.IV: 13673804 - 13677886 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070242.2 |
NP_502643.1 |
6 - 7 |
1207 |
141 |
View Full Product Details