Assay Design
Gene Transcripts
- Gene Symbol
- sas-6
- Entrez Gene ID
-
178345
- Gene Name
- Spindle assembly abnormal protein 6
- Gene Aliases
-
CELE_Y45F10D.9
- Chromosome Location
-
Chr.IV: 13786945 - 13788731 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070259.6 |
NP_502660.1 |
3 - 4 |
1359 |
108 |
View Full Product Details