Assay Design



Gene Transcripts

Gene Symbol
ssu-2
Entrez Gene ID
178379
Gene Name
protein-coding
Gene Aliases
CELE_Y67A10A.6
Chromosome Location
Chr.IV: 14361247 - 14368322 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268875.3 NP_001255804.1 1 - 2 76 86
NM_001268876.2 NP_001255805.1 1 - 2 81 86

View Full Product Details