Assay Design



Gene Transcripts

Gene Symbol
fcd-2
Entrez Gene ID
178418
Gene Name
protein-coding
Gene Aliases
CELE_Y41E3.9
Chromosome Location
Chr.IV: 15025706 - 15037036 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268919.2 NP_001255848.1 8 - 9 3460 125
NM_001268920.3 NP_001255849.1 4 - 5 1681 125
NM_001268921.3 NP_001255850.1 2 - 3 610 125

View Full Product Details