Assay Design
Gene Transcripts
- Gene Symbol
- srt-48
- Entrez Gene ID
-
189835
- Gene Name
- Serpentine Receptor, class T
- Gene Aliases
-
CELE_Y41E3.15
- Chromosome Location
-
Chr.IV: 15065866 - 15068918 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070420.3 |
NP_502821.3 |
2 - 3 |
341 |
90 |
View Full Product Details