Assay Design
Gene Transcripts
- Gene Symbol
- sra-30
- Entrez Gene ID
-
189805
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_Y40H7A.6
- Chromosome Location
-
Chr.IV: 15192299 - 15203023 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070432.3 |
NP_502833.3 |
6 - 7 |
885 |
123 |
View Full Product Details