Assay Design
Gene Transcripts
- Gene Symbol
- lgc-52
- Entrez Gene ID
-
190669
- Gene Name
- Ligand-Gated ion Channel
- Gene Aliases
-
CELE_Y73F8A.2
- Chromosome Location
-
Chr.IV: 15236123 - 15244067 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_070438.4 |
NP_502839.3 |
4 - 5 |
830 |
119 |
View Full Product Details